Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Crizotinib
GREENER_CHOICESDP

Catalog No. 436850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-685-0 50 mg
43-681-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-685-0 Supplier R&D Systems™ Supplier No. 4368/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent c-MET/ALK inhibitor

Potent inhibitor of c-MET and anaplastic lymphoma kinase (ALK) (cell IC50 values are 8.0 and 20 nM respectively). Selective for c-MET and ALK against >120 different kinases. Displays antitumor efficacy in multiple tumor models; inhibits c-MET-dependent proliferation, migration and invasion of human tumor cells in vitro. Orally bioavailable.

Chemical Identifiers

CAS 877399-52-5
Molecular Formula C21H22Cl2FN5O

Specifications

Assay Percent Range >99%
Quantity 50 mg
Product Line Tocris
Formula Weight 450.34
Molecular Weight (g/mol) 450.34
Chemical Name or Material Crizotinib
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.