Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crk Antibody (3G11E8), Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
Antigen | Crk |
---|---|
Clone | 3G11E8 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Crk Monoclonal specifically detects Crk in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Crk | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
adapter molecule crk, avian sarcoma virus CT10 (v-crk) oncogene homolog, CRKII, p38, Proto-oncogene c-Crk, v-crk avian sarcoma virus CT10 oncogene homolog, v-crk sarcoma virus CT10 oncogene homolog (avian) | |
CRK | |
IgG2b | |
Unpurified |
3G11E8 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Ovarian Carcinoma Cell Markers | |
Ascites with 0.03% Sodium Azide | |
1398 | |
Purified recombinant fragment of human CRK expressed in E. Coli. | |
Primary | |
42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title