Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crk Antibody (SR1635), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322261100UL
Description
Crk Monoclonal antibody specifically detects Crk in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
Crk | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
SR1635 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
adapter molecule crk, avian sarcoma virus CT10 (v-crk) oncogene homolog, CRKII, p38, Proto-oncogene c-Crk, v-crk avian sarcoma virus CT10 oncogene homolog, v-crk sarcoma virus CT10 oncogene homolog (avian) | |
A synthesized peptide derived from human Crk (Uniprot #: P46108) | |
100 μg | |
Apoptosis, Ovarian Carcinoma Cell Markers, Phospho Specific | |
1398 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction