Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CrkL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581877 Supplier Invitrogen™ Supplier No. PA581877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEYVRTLYDF PGNDAEDLPF KKGEILVIIE KPEEQWWSAR NKDGRVGMIP VPYVEKLVRS SPHGKHGNRN SNSYGIPEPA HAYAQPQTTT PLPAVSGSPG AAITPLPSTQ NGPVFAKAIQ KRVPCAYDKT ALALEVGDIV KVTRMNINGQ WEGEVNGRKG LFPFTHVKIF DPQNPDE Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.

The CRKL gene encodes the CRK-like protein, an adaptor protein that plays a crucial role in intracellular signaling pathways related to cell proliferation, differentiation, and migration. CRKL contains SH2 and SH3 domains that facilitate interactions with other signaling molecules, influencing multiple pathways such as the MAPK, PI3K, and STAT pathways. It is involved in processes like cytoskeleton reorganization and immune cell signaling, with significant implications for development and cancer. CRKL is notably associated with Philadelphia chromosome-positive leukemias, where it interacts with the BCR-ABL fusion protein, contributing to leukemogenesis. Additionally, dysregulation of CRKL can affect cardiovascular development, leading to congenital heart defects. Research into CRKL focuses on understanding its role in signal transduction and its potential as a therapeutic target due to its involvement in oncogenic and developmental pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CrkL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRKL
Gene Accession No. P46109
Gene Alias 1110025F07Rik; AA589403; AI325100; CRK like proto-oncogene, adaptor protein; CRKL; crkl {ECO:0000312; crk-like protein; crk-like protein {ECO:0000250; Crkol; EMBL:AAH85865.1, ECO:0000312; OTTHUMP00000198539; RGD:1308531}; UniProtKB:P46109}; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk sarcoma virus CT10 oncogene homolog-like
Gene Symbols CRKL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRKL. Recombinant protein control fragment (Product #RP-91386).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1399
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.