Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRKL, Rabbit anti-Human, PE, Clone: G4, Abnova™

Catalog No. 89335716 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-716 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-716 Supplier Abnova Corporation Supplier No. MAB18882
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen CRKL
Applications Flow Cytometry
Classification Monoclonal
Clone G4
Conjugate PE
Dilution Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Gene v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Gene Alias -
Gene Symbols CRKL
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y207 of human CRKL.
Quantity 10 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1399
Target Species Human, Mouse
Content And Storage Store at 4°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.