Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CrkRS Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10087011B
Description
CrkRS Polyclonal antibody specifically detects CrkRS in Human samples. It is validated for Western BlotSpecifications
CrkRS | |
Polyclonal | |
Western Blot | |
Cdc2-related kinase, arginine/serine-richCrkRS, Cell division cycle 2-related protein kinase 7, Cell division protein kinase 12, CRK7CDC2-related protein kinase 7, CRKR, CRKRS, cyclin-dependent kinase 12, EC 2.7.11, EC 2.7.11.23, hCDK12, KIAA0904EC 2.7.11.22 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cell division cycle 2 (CDC2)-related kinase arginine/serine-rich using the numbering given in entry NP_057591.2 (GeneID 51755). | |
0.1 mL | |
Protein Kinase | |
51755 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction