Learn More
Invitrogen™ CRLF1 Polyclonal Antibody

Description
Predicted to react with Human and Mouse samples.
Specifications
Specifications
| Antigen | CRLF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CRLF1 |
| Gene Accession No. | O75462, Q9JM58 |
| Gene Alias | CISS; CISS1; class I cytokine receptor; CLF; CLF-1; CRLF1; CRLM3; CRLM-3; cytokine receptor like factor 1; cytokine receptor like molecule 3; cytokine receptor-like factor 1; Cytokine receptor-like molecule 3; cytokine type 1 receptor CRLP-1; cytokine-like factor 1; Novel cytokine receptor 6; NR6; NR6.1; UNQ288/PRO327; ZcytoR5 |
| Gene Symbols | CRLF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.