Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRLF1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988697
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8697 Supplier Bioss Supplier No. BS8663R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRLF1 encodes a member of the cytokine type I receptor family. The protein forms a secreted complex with cardiotrophin-like cytokine factor 1 and acts on cells expressing ciliary neurotrophic factor receptors. The complex can promote survival of neuronal cells. Mutations in this gene result in Crisponi syndrome and cold-induced sweating syndrome.

Specifications

Antigen CRLF1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CRLF1
Gene Accession No. O75462
Gene Alias CISS; CISS1; class I cytokine receptor; CLF; CLF-1; CRLF1; Crlm3; CRLM-3; cytokine receptor like factor 1; cytokine receptor like molecule 3; Cytokine receptor-like factor 1; Cytokine receptor-like molecule 3; cytokine type 1 receptor CRLP-1; Cytokine-like factor 1; Novel cytokine receptor 6; NR6; NR6.1; UNQ288/PRO327; ZcytoR5
Gene Symbols CRLF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRLF1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9244
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.