Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRLF3 Monoclonal Antibody (OTI2A10), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene CF812346

Encompass
Encompass_Preferred

Catalog No. 50-254-3700


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

CRLF3 Monoclonal antibody specifically detects CRLF3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

CRLF3
Monoclonal
See Label
PBS with 8% trehalose and no preservative; pH 7.3
P30305
Mouse
Protein A/G
RUO
994
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2A10
Unconjugated
CDC25B
BB164954; Creme9; CREME-9; CRLF3; CRLM9; cytokine receptor like factor 3; cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; cytokine receptor-related factor 4; Cytokine receptor-related protein 4; CYTOR4; FRWS; P48; p48.2; Type I cytokine receptor-like factor p48
Human recombinant protein fragment corresponding to amino acids 421-567 of human CDC25B (NP_004349) produced in E.coli.
100 μg
Primary
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.