Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRLF3 Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene
SDP

Catalog No. 502543698
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3698 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3698 Supplier OriGene Supplier No. CF812348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRLF3 Monoclonal antibody specifically detects CRLF3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CRLF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2H3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CRLF3
Gene Accession No. Q8IUI8, Q9Z2L7
Gene Alias BB164954; Creme9; CREME-9; CRLF3; CRLM9; cytokine receptor like factor 3; cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; cytokine receptor-related factor 4; Cytokine receptor-related protein 4; CYTOR4; FRWS; P48; p48.2; Type I cytokine receptor-like factor p48
Host Species Mouse
Immunogen Full length human recombinant protein of human CRLF3 (NP_057070) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51379, 54394, 54395
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.