Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRLF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103401 Supplier Invitrogen™ Supplier No. PA5103401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CRLF3.

May play a role in the negative regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRLF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CRLF3
Gene Accession No. Q8IUI8, Q9Z2L7
Gene Alias BB164954; Creme9; CREME-9; CRLF3; CRLM9; cytokine receptor like factor 3; cytokine receptor-like factor 3; Cytokine receptor-like molecule 9; cytokine receptor-related factor 4; Cytokine receptor-related protein 4; CYTOR4; FRWS; P48; p48.2; Type I cytokine receptor-like factor p48
Gene Symbols CRLF3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CRLF3(Accession Q8IUI8), corresponding to amino acid residues G302-V352.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51379, 54394, 54395
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.