Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRLS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525338
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525338 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525338 Supplier Thermo Scientific Supplier No. PA525338
Add to Cart
Add to Cart

CRLS1 Polyclonal Antibody

CRLS1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CRLS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CRLS1
Gene Accession No. Q9UJA2
Gene Alias 0610009I22Rik; 4930557M15Rik; 5730490M08Rik; C20orf155; cardiolipin synthase; cardiolipin synthase (CMP-forming); cardiolipin synthase 1; cardiolipin synthetase; CLS; CLS1; CRLS1; dJ967N21.6; GCD10; Protein GCD10 homolog; RGD1311037
Gene Symbols CRLS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 198-226 amino acids from the C-terminal region of human CRLS1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.