Learn More
CRM1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | CRM1 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1560
![]() |
Bethyl Laboratories, Inc
IHC00139T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1559
![]() |
Bethyl Laboratories, Inc
IHC00139 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque and Wh
The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.Specifications
CRM1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Chromosome Region Maintece 1 | |
Between 1025 and C-term | |
Primary | |
4° C | |
XPO1 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O14980, Q6P5F9 | |
103573, 7514 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.