Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRM1 Mouse anti-Human, Mouse, Rat, Clone: 2D3B6, Proteintech
SDP

Catalog No. 501737169 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7169 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7169 Supplier Proteintech Group Inc Supplier No. 667631IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene mediates leucine-rich nuclear export signal (NES)-dependent protein transport. Exportin 1 specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.

Specifications

Antigen CRM1
Applications Western Blot
Classification Monoclonal
Clone 2D3B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene XPO1
Gene Accession No. O14980, Q6P5F9, Q80U96
Gene Alias CRM1, DKFZp686B1823, Exp1, Exportin 1, XPO1
Gene Symbols Xpo1
Host Species Mouse
Immunogen CRM1 Fusion Protein Ag27337
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103573, 7514, 85252
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.