Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRM1 Rabbit anti-Human, Mouse, Rat, Clone: 1J0L10, Novus Biologicals™
SDP

Catalog No. p-7247321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166742 20 μg
NB166741 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166742 Supplier Novus Biologicals Supplier No. NBP31582320UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CRM1 Monoclonal antibody specifically detects CRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CRM1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1J0L10
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Chromosome region maintenance 1 protein homolog, DKFZp686B1823, emb, exp1, exportin 1 (CRM1 homolog, yeast), exportin 1 (CRM1, yeast, homolog), exportin-1, Exportin-1 (required for chromosome region maintenance), yeast, homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 972-1071 of human CRM1 (O14980).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7514
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.