Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRMP2 Antibody (20D8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CRMP2 |
---|---|
Clone | 20D8 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
CRMP2 Monoclonal antibody specifically detects CRMP2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
CRMP2 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
A synthesized peptide derived from Human CRMP2 [UniProt Q16555] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
20D8 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1808 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title