Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRMP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323261
Description
CRMP2 Polyclonal antibody specifically detects CRMP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
CRMP2 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:100 | |
Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
CRMP2 synthetic peptide (coupled to carrier protein) corresponding to amino acids in the C-terminal region of human CRMP2. This sequence is conserved in rat and mouse CRMP2, and has high homology to the conserved site in CRMP1. It has 50% homology to CRMP3 and CRMP5. [UniProt# Q16555] | |
0.1 mL | |
Neuroscience, Phospho Specific | |
1808 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction