Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRMP2, p Ser522 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323262
Description
CRMP2 Polyclonal antibody specifically detects CRMP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CRMP2 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
Phospho-CRMP2 (Ser-522) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-522 in human CRMP2. This sequence is conserved in rat and mouse CRMP2, and has homology to the conserved site in CRMP1 and CRMP4, but is not conserved in CRMP3 and CRMP5. [UniProt# Q16555] | |
0.1 mL | |
Neuroscience, Phospho Specific | |
1808 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction