Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRMP2 Rabbit anti-Human, Mouse, Rat, Clone: 0A1T1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CRMP2 |
---|---|
Clone | 0A1T1 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CRMP2 Monoclonal antibody specifically detects CRMP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
CRMP2 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1808 | |
IgG | |
Affinity purified |
0A1T1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Collapsin response mediator protein 2, CRMP-2, CRMP2N2A3, DHPRP2, dihydropyrimidinase-like 2, dihydropyrimidinase-like 2 long form, dihydropyrimidinase-related protein 2, DRP2collapsin response mediator protein hCRMP-2, DRP-2ULIP-2, ULIP2, Unc-33-like phosphoprotein 2 | |
A synthetic peptide corresponding to a sequence within amino acids 473-572 of human CRMP2 (Q16555). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title