Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536585 Supplier Invitrogen™ Supplier No. PA536585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 50 kDa. Purity is >95% by SDS-PAGE.

Collapsin response mediator proteins (CRMPs), including CRMP-1 (DRP-1), CRMP-2 (DRP-2 or TOAD64), CRMP-3 (DRP-4), CRMP-4 (DRP-3) and CRMP-5 (DRP-5), mediate signal transduction after exposure of neural cells to the axon guidance molecule Semaphorin 3A (SEMA3A)/collapsin. CRMPs are present in the developing cerebral cortex and neocortical neurons and are responsive to SEMA3A. In the adult brain, the expression of CRMPs is dramatically downregulated. However, they remain expressed in structures that retain their capacity for differentiation and plasticity. CRMP-3 is expressed transiently in developing spinal cord and selectively in the postnatal cerebellum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.2
Gene DPYSL4
Gene Accession No. O14531, O35098, Q62951
Gene Alias AI173505; Collapsin response mediator protein 3; CRMP3; CRMP-3; dihydropyrimidinase like 4; dihydropyrimidinase-like 4; dihydropyrimidinase-related protein 4; DPY4; Dpys4; DPYSL4; DRP-4; ULIP4; ULIP-4; ULIP4 protein; unc-33-like phosphoprotein 4; UNC33-like phosphoprotein 4
Gene Symbols DPYSL4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 111-160 of human CRMP-3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10570, 25417, 26757
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.