Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRMP3 Rabbit anti-Feline, Human, Mouse, Non-human Primate, Porcine, Rat, Polyclonal, FabGennix

Catalog No. 502206793
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6793 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6793 Supplier Fabgennix Inc Supplier No. DRP4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRMP3 Polyclonal antibody specifically detects CRMP3 in Feline, Human, Mouse, Non-human Primate, Porcine, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen CRMP3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DPYSL4
Gene Accession No. O14531, O35098, Q62951
Gene Alias AI173505; Collapsin response mediator protein 3; CRMP3; CRMP-3; dihydropyrimidinase like 4; dihydropyrimidinase-like 4; dihydropyrimidinase-related protein 4; DPY4; Dpys4; DPYSL4; DRP-4; ULIP4; ULIP-4; ULIP4 protein; unc-33-like phosphoprotein 4; UNC33-like phosphoprotein 4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 509-527 of Mouse CRMP3. The peptide was covalently modified to achieve the desired antigenicity before coupling to carrier protein.Sequence:PAKPGSGTQARASCSGKIS
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100512868, 101086601, 10570, 25417, 26757, 450833
Target Species Feline, Human, Mouse, Non-human Primate, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.