Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRMP5 Monoclonal Antibody (1503CT789.209.250.52)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537679
Description
CRMP5 Monoclonal Antibody for Western Blot, ICC/IF
The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
Specifications
CRMP5 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9BPU6, Q9EQF6, Q9JHU0 | |
DPYSL5 | |
recombinant protein of human DPYL5. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Western Blot, Immunocytochemistry | |
1503CT789.209.250.52 | |
Unconjugated | |
DPYSL5 | |
collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6 | |
Mouse | |
Protein G | |
RUO | |
56896, 65208, 65254 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction