Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRMP5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104345
Description
Antibody detects endogenous levels of total CRMP5.
The collapsin response mediator protein (CRMP) family of proteins (also referred to as ULIP6) are a group of neurologic proteins that have been found to play key roles in growth cone guidance during neural development. CRMP5 has relatively low sequence homology with the other four members of the CRMP family. CRMP5 is expressed in the developing nervous system in an expression pattern that resembles CRMP2 and has a peak expression during the first postnatal week. Elevated levels of autoantibody to CRMP5 have been shown to be linked to paraneoplastic autoimmune optic neuritis and other paraneoplastic neoplasms. Auto-immunity against CRMP5 expression (predominantly the N-terminal epitopes) is also being investigated as an oncological marker in the respiratory system and in the thymus.
Specifications
CRMP5 | |
Polyclonal | |
Unconjugated | |
DPYSL5 | |
collapsin response mediator protein 5; collapsin response mediator protein-5; CRAM; CRMP3-associated molecule; Crmp5; CRMP-5; dihydropyrimidinase like 5; dihydropyrimidinase-like 5; dihydropyrimidinase-related protein 5; Dpysl5; DRP-5; Ulip6; ULIP-6; ULIP6 protein; UNC33-like phosphoprotein 6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
56896, 65208, 65254 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BPU6, Q9EQF6, Q9JHU0 | |
DPYSL5 | |
A synthesized peptide derived from human DPYSL5(Accession Q9BPU6), corresponding to amino acid residues V519-W564. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction