Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRNKL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3186 Supplier Bioss Supplier No. BS14065R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRNKL1 is a protein coding gene.

Specifications

Antigen CRNKL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CRNKL1
Gene Accession No. Q9BZJ0
Gene Alias 1200013P10Rik; 5730590A01Rik; BOS_12978; C80326; CGI-201; CLF; Clf1; Crn; Crn, crooked neck-like 1; Crn, crooked neck-like 1 (Drosophila); CRNKL1; Crooked neck homolog; crooked neck pre-mRNA splicing factor 1; crooked neck pre-mRNA splicing factor-like 1; crooked neck pre-mRNA splicing factor-like 1 (Drosophila); Crooked neck protein; crooked neck-like 1; Crooked neck-like protein 1; crooked neck-like protein 1-like; hCrn; MSTP021; SYF3; SYF3 pre-mRNA-splicing factor
Gene Symbols CRNKL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRNKL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51340
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.