Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CROCC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554346 Supplier Invitrogen™ Supplier No. PA554346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKRLEKQNLE KDQVNKDLTE KLEALESLRL QEQAALETED GEGLQQTLRD LAQAVLSDSE SGVQLSGSER TADASNGS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

Major structural component of the ciliary rootlet, a cytoskeletal-like structure in ciliated cells which originates from the basal body at the proximal end of a cilium and extends proximally toward the cell nucleus. Furthermore, is required for the correct positioning of the cilium basal body relative to the cell nucleus, to allow for ciliogenesis (PubMed:27623382). Contributes to centrosome cohesion before mitosis (PubMed:16203858). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CROCC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CROCC
Gene Accession No. Q5TZA2
Gene Alias ciliary rootlet coiled-coil protein; ciliary rootlet coiled-coil, rootletin; CROCC; KIAA0445; ROLT; rootletin; rootletin, ciliary rootlet protein; Tax1-binding protein 2
Gene Symbols CROCC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CROCC. Recombinant protein control fragment (Product #RP-93571).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.