Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crocetin, free acid, >90%, MP Biomedicals™

Catalog No. ICN19354305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19354305 5 mg
ICN19354301 1 mg
ICN19354325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19354305 Supplier MP Biomedicals, Inc Supplier No. 0219354305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27876-94-4
Molecular Formula C20H24O4
Molecular Weight (g/mol) 328.408
InChI Key PANKHBYNKQNAHN-MQQNZMFNSA-N
Synonym crocetin, trans-crocetin, 8,8'-diapocarotenedioic acid, natural yellow 6, 8,8'-diapocarotene-8,8'-dioic acid, 8,8'-diapo-psi,psi-carotenedioic acid, ccris 7484, unii-20tc155l9c, 2e,4e,6e,8e,10e,12e,14e-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid, .alpha.-crocetin
PubChem CID 5281232
ChEBI CHEBI:3918
IUPAC Name (2E,4E,6E,8E,10E,12E,14E)-2,6,11,15-tetramethylhexadeca-2,4,6,8,10,12,14-heptaenedioic acid
SMILES CC(=CC=CC=C(C)C=CC=C(C)C(=O)O)C=CC=C(C)C(=O)O

Specifications

Color Orange
Assay Percent Range >90%
Quantity 5 mg
Formula Weight 328.4
Physical Form Powder
Chemical Name or Material Crocetin, free acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.