Learn More
Croconic acid, 98%
Description
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
Solubility
Soluble in water and ethanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼280°C (decomposition) |
| Odor | Odorless |
| Quantity | 1 g |
| Solubility Information | Soluble in water and ethanol. |
| Formula Weight | 142.07 |
| Percent Purity | 98% |
| Chemical Name or Material | Croconic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.