Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CROP Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074625AF405
Description
CROP Polyclonal antibody specifically detects CROP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CROP | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, Cisplatin resistance-associated-overexpressed protein, CREAP1, CREAP-1LUC7ACRA, CRE-associated protein 1, CROPFLJ11063, hLuc7A, Luc7A, LUC7-like 3 (S. cerevisiae), luc7-like protein 3, O48, OA48-18, Okadaic acid-inducible phosphoprotein OA48-18 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cisplatin resistance-associated overexpressed protein using the numbering given in entry NP_057508.2 (GeneID 51747). | |
0.1 mL | |
Apoptosis | |
51747 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction