Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CROP/Luc7A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572379
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2379 100 μL
50-157-2380 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2379 Supplier Bethyl Laboratories, Inc Supplier No. IHC00600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Bovine, Dog, Rabbit, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Nile tilap

The function of this protein remains unknown.

Specifications

Antigen CROP/Luc7A
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene LUC7L3
Gene Accession No. O95232, Q5SUF2
Gene Alias cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, cisplatin resistance-associated-overexpressed protein, CRA, CREAP-1, CRE-associated protein, CRE-associated protein 1, CROP, FLJ11063, hLuc7A, LUC7A, LUC7-like 3 (S. cerevisiae), LUC7-like 3 pre-mRNA splicing factor, OA48-18, okadaic acid-inducible phosphoprotein OA48-18
Gene Symbols Luc7l3
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51747, 67684
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.