Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croscarmellose Sodium, NF, Spectrum™ Chemical
SDP

Catalog No. 18601881 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-881 25 kg Fiber Drum
18-601-879 500 g Poly Bottle
18-601-880 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-881 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C136625KGBL
Only null left
Add to Cart
Add to Cart

C1366, 74811-65-7

Spectrum™ Chemical Croscarmellose Sodium, NF is used commonly as a FDA-approved disintegrant in pharmaceutical manufacturing. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 74811-65-7

Specifications

CAS Max % 100%
Ignition Residue 14.0-28%
Loss on Drying 10%
Packaging Fiber Drum
Quantity 25 kg
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.