Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cross-Reactive CD49d Rat anti-nan, BV711, Clone: PS/2 (also known as PS-2), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756083
Description
Cross-Reactive CD49d Monoclonal antibody specifically detects Cross-Reactive CD49d in nan samples. It is validated for Flow cytometry
Specifications
Cross-Reactive CD49d | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
3676 | |
Liquid |
Flow Cytometry | |
PS/2 (also known as PS-2) | |
Brilliant Violet 711 | |
ITGA4; Integrin alpha 4; Itga4; LPAM alpha; VLA-4 alpha; α4 Integrin | |
Cells from the Mouse P815 Mastocytoma Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction