Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CROT Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537981
Description
This antibody is predicted to react with rat and dog based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
Specifications
| CROT | |
| Polyclonal | |
| Unconjugated | |
| Crot | |
| 1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 54677, 74114 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9DC50, Q9UKG9 | |
| Crot | |
| Peptide with sequence NYFKTESEGHCPTH, from the internal region of rat Crot (aa178-199). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction