Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CROT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553985 Supplier Invitrogen™ Supplier No. PA553985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVPVHKVGNT PLDMNQFRML FSTCKVPGIT RDSIMNYFRT ESEGRSPNHI VVLCRGRAFV FDVIHEGCLV TPPELLRQLT YIHKKCHSEP DGPGIAALTS EERTRWAKAR EYLIGLD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CROT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Crot
Gene Accession No. Q9UKG9
Gene Alias 1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643
Gene Symbols Crot
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CROT. Recombinant protein control fragment (Product #RP-92590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54677
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.