Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CROT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553906
Description
Immunogen sequence: LAKSTEERTF QYQDSLPSLP VPSLEESLKK YLESVKPFAN QEEYKKTEEI VQKFQSGIGE KLHQKLLERA KGKRNWLEEW WLNVAYLDVR IPSQLNVNFA GPAAHFEHYW PPKEGTQLER GSITLWH Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
CROT encodes a member of the carnitine/choline acetyltransferase family. The encoded protein converts 4, 8-dimethylnonanoyl-CoA to its corresponding carnitine ester. This transesterification occurs in the peroxisome and is necessary for transport of medium- and long- chain acyl-CoA molecules out of the peroxisome to the cytosol and mitochondria. The protein thus plays a role in lipid metabolism and fatty acid beta-oxidation. Alternatively spliced transcript variants have been described.
Specifications
| CROT | |
| Polyclonal | |
| Unconjugated | |
| Crot | |
| 1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKG9 | |
| Crot | |
| Recombinant protein corresponding to Human CROT. Recombinant protein control fragment (Product #RP-92589). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction