Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Croton Oil, Spectrum™ Chemical

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Steel Pail
Description
Spectrum™ Chemical Croton Oil is an oil prepared from the seeds of the tree, Croton tiglium. Croton oil is the source of the organic compound phorbol and is the basis of rejuvenating chemical peels, due to the caustic exfoliating effects it has on the dermal components of the skin. Used in conjunction with phenol solutions, it results in an intense reaction which leads to initial skin sloughing and then eventual regeneration. In traditional Chinese medicine it is used as an ingredient in some liniments.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Refractive Index | 1.4760-1.4780 |
Quantity | 1 kg |
UN Number | UN2810 |
Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction