Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Crotonaldehyde Monoclonal Antibody (2A8.1), FITC
GREENER_CHOICE

Catalog No. PIMA545850
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545850 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545850 Supplier Invitrogen™ Supplier No. MA545850
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45850 was sufficient for detection of Crotonaldehyde-modified proteins in 1 μg of Crotonaldehyde-BSA conjugate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for Crotonaldehyde modified proteins. Does not cross-react with Acrolein, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.

Crotonaldehyde is a chemical compound that is an unsaturated aldehyde, used as a preservative and found in food such as soybean oil.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Crotonaldehyde
Applications Dot Blot, ELISA, Western Blot
Classification Monoclonal
Clone 2A8.1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
Gene Alias CRA; CRA-lysine adduct; Croton aldehyde; Crotonaldehyde
Host Species Mouse
Immunogen Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.