Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonaldehyde Mouse, ATTO 390, Clone: 2A8.1, Abnova™

Catalog No. 89339044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-044 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-044 Supplier Abnova Corporation Supplier No. MAB19759
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Crotonaldehyde Monoclonal antibody specifically detects Crotonaldehyde in samples. It is validated for WB, ELISA.

Specifications

Antigen Crotonaldehyde
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2A8.1
Conjugate Atto 390
Host Species Mouse
Immunogen Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.