Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonaldehyde, predominantly trans, 98+%

Catalog No. aal03362ad
Click to view available options
Quantity:
50 mL
250 mL

CAS: 4170-30-3 | C4H6O | 70.091 g/mol

Crotonaldehyde is used as intermediates. It is an excellent prochiral dienophile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crotonaldehyde is used as intermediates. It is an excellent prochiral dienophile.

Solubility
Soluble in water. (150g/L) at 20°C.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with bases, heat, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4170-30-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00007003
InChI Key MLUCVPSAIODCQM-NSCUHMNNSA-N
Synonym crotonaldehyde, 2-butenal, crotylaldehyde, crotonal, 2e-but-2-enal, trans-crotonaldehyde, e-crotonaldehyde, crotonic aldehyde, 1-formylpropene, trans-2-butenal
PubChem CID 447466
ChEBI CHEBI:41607
IUPAC Name (E)-but-2-enal
SMILES CC=CC=O

Specifications

Melting Point -76°C
Density 0.848
Boiling Point 103°C to 104°C
Flash Point 8°C (46°F)
Odor Pungent
Refractive Index 1.437
Quantity 50 mL
UN Number UN1143
Beilstein 906731
Sensitivity Air sensitive
Merck Index 14,2596
Solubility Information Soluble in water. (150g/L) at 20°C.
Formula Weight 70.09
Percent Purity ≥98%
Chemical Name or Material Crotonaldehyde, predominantly trans
Show More Show Less
Hazard Category H225-H301-H310+H330-H315-H318-H335-H341-H372
Hazard Statement GHS H Statement
H225-H301-H311-H330-H341-H373-H315-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CROTONALDEHYDE, STABILIZED
EINECSNumber 224-030-0
RTECSNumber GP9499000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.