Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonic acid, 98%

Catalog No. p-94119
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 107-93-7 | C4H6O2 | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-93-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002701
InChI Key LDHQCZJRKDOVOX-NSCUHMNNSA-N
Synonym crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
PubChem CID 637090
ChEBI CHEBI:41131
IUPAC Name (E)-but-2-enoic acid
SMILES CC=CC(=O)O

Specifications

Melting Point 72.0°C
Color White to Yellow
Boiling Point 185.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3CH=CHCO2H
Quantity 500 g
Beilstein 02, IV, 1491
Merck Index 15, 2588
Solubility Information Solubility in water: 94g/L (25°C). Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 86.09
Percent Purity 98%
Physical Form Crystalline Flakes
Chemical Name or Material Crotonic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 203-533-9
RTECSNumber GQ2900000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.