Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonic Acid 98.0+%, TCI America™
SDP

Catalog No. C0416500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0416500G 500 g
C041625G 25 g
C0416100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0416500G Supplier TCI America Supplier No. C0416500G

Chemical Identifiers

CAS 107-93-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002701
InChI Key LDHQCZJRKDOVOX-NSCUHMNNSA-N
Synonym crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
PubChem CID 637090
ChEBI CHEBI:41131
IUPAC Name (E)-but-2-enoic acid
SMILES CC=CC(=O)O

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 185°C
Quantity 500 g
UN Number 2823
Formula Weight 86.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Crotonic Acid
EINECSNumber (2)-0963
RTECSNumber GQ2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.