Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Crotonyl-HIST1H2BC (Lys20) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117476 Supplier Invitrogen™ Supplier No. PA5117476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. The protein has antibacterial and antifungal antimicrobial activity. The main transcript variant of this gene is intronless and encodes a replication-dependent histone that is a member of the histone H2B family. This transcript variant lacks a polyA tail but instead contains a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Crotonyl-HIST1H2BC (Lys20)
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.161 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene H2BC5
Gene Accession No. P58876, P62807
Gene Alias dJ221C16.3; H2B Clustered Histone 4; H2B histone family, member L; H2B.1; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bc; histone cluster 1, H2bc; histone H2B type 1-C/E/F/G/I; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l
Gene Symbols H2BC5, Hist1h2bc
Host Species Rabbit
Immunogen Peptide sequence around site of Crotonyl-Lys (20) derived from Human Histone H2B type 1-C/E/F/G/I.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3017
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.