Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Crotonyl-Histone H1.2 (Lys158) Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144318
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144318 Supplier Invitrogen™ Supplier No. PA5144318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Crotonyl-Histone H1.2 (Lys158) Polyclonal Antibody for ICC/IF, ELISA, ChIP

Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Crotonyl-Histone H1.2 (Lys158)
Applications ChIP Assay, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene H1-2
Gene Accession No. P16403
Gene Alias 0610008C09Rik; CTL-1; H1 histone family, member 2; H1 VAR.1; H1.2; H1.2 linker histone, cluster member; H1-2; H1C; H1f2; H1s-1; H1var1; His1a; Hist1h1c; histone 1, H1c; histone cluster 1, H1c; histone H1; histone H1.1; histone H1.2; Histone H1c; histone H1d; Histone H1s-1; LOC684681; MGC3992; similar to Histone H1.2 (H1 VAR.1) (H1c)
Gene Symbols H1-2, HIST1H1C
Host Species Rabbit
Immunogen Peptide sequence around site of Crotonyl-Lys (158) derived from Human Histone H1.2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3006
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.