Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Crovalimab Chimeric Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542180
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Crovalimab, a Sequential Monoclonal Antibody Recycling Technology (SMART) antibody was engineered for extended self-administered subcutaneous dosing of small volumes in diseases amenable for C5 inhibition.
Specifications
Crovalimab Chimeric | |
Recombinant Monoclonal | |
Unconjugated | |
RG-6107; RO7112689; SKY-59 | |
Human C5. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
7.13 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction