Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRP Chicken anti-Rat, Polyclonal , Abnova™

Catalog No. 89933999 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-999 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-999 Supplier Abnova Corporation Supplier No. PAB29703

Chicken polyclonal antibody raised against rat Crp.

Specifications

Antigen CRP
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene Alias Aa1249, Ab1341, Ab2196, Ac1114, Ac1262, Ac2069, Ba2693
Gene Symbols Crp
Host Species Chicken
Immunogen Native purified rat Crp.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25419
Target Species Rat
Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.