Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRP Monoclonal Antibody (C11F2), Cyanine7, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM0391MCY7

This item is not returnable.
View return policy
Description
CRP Monoclonal antibody specifically detects CRP in Human, Mouse, Rat samples. It is validated for ImmunocytochemistrySpecifications
CRP | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P02741, P14847, P48199 | |
Mouse | |
Protein A | |
RUO | |
12944, 1401, 25419 | |
-20°C or -80°C if preferred | |
Liquid |
Immunocytochemistry | |
C11F2 | |
Cyanine7 | |
Crp | |
Aa1249; Ab1-341; Ab2-196; Ac1-114; Ac1262; Ac2-069; acute phase protein; CRP; AI255847; Ba2-693; c reactive; C-reactive preprotein; C-reactive protein; C-reactive protein member of the pentraxin family; C-reactive protein(1-205); C-reactive protein, member of the pentraxin family; C-reactive protein, pentraxin-related; C-reactive protein, petaxin related; CRP; CRP1; CSRP; CYRP; D1S181E; DKFZp686M148; DKFZp686M149; MGC149895; MGC88244; pentraxin 1; PTX1; RP11-419N10.4 | |
Recombinded CRP protein (amino acids 19-224). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction