Learn More
CRP Polyclonal Antibody, FITC, Rockland™

Goat Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | CRP |
---|---|
Concentration | 1 mg/mL |
Applications | Dot Blot, ELISA, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1342
![]() ![]() |
Rockland Immunochemicals
20912320100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-1343
![]() ![]() |
Rockland Immunochemicals
2091232S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-fluorescein, anti-Goat Serum, purified and partially purified Human CRP.
CRP (C-reactive protein) is a major cyclic, pentameric acute phase protein compound consisting of five identical, noncovalently bound, nonglycosylated subunits. Eeach subunit is made up of 206 amino acids, and has a molecular weight of 24 kDa. CRP is produced by the liver, and its plasma levels rise dramatically during inflammatory processes occurring in the body. CRP is an initiator of classical complement cascade, binds to several nuclear components (chromatin, histones, etc.), plays an important role in innate immunity, and mediates activities associated with pre-immune nonspecific host resistance. Specifically, there is a 100-1000 fold increase of CRP in response to injury, infection or inflammation. CRP is opsonic, an initiator of the classical complement cascade, and an activator of monocytes/macrophages. CRP also binds to several nuclear components including chromatin, histones and snRNP, suggesting that it may play a role as a scavenger during cell necrosis. CRP is involved in several host defense related functions based on its ability to recognize foreign pathogens and damaged cells of the host, and initiates pathogen elimination by interacting with humoral and cellular effector systems in the blood. Patients with elevated basal levels of CRP have been shown to be an increased risk for hypertension and cardiovascular disease.Specifications
CRP | |
Dot Blot, ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
FITC | |
Goat | |
Human | |
P02741 | |
1401 | |
Human CRP (Capsular Reactive Protein) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Crp |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.01M sodium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% thimerosal; pH 7.2 | |
Aa1249; Ab1-341; Ab2-196; Ac1-114; Ac1262; Ac2-069; AI255847; Ba2-693; C-reactive protein; C-reactive protein member of the pentraxin family; C-reactive protein(1-205); C-reactive protein, member of the pentraxin family; C-reactive protein, pentraxin-related; C-reactive protein, petaxin related; C-reactive protein-like; CRP; CRP1; CSRP; CYRP; D1S181E; DKFZp686M148; DKFZp686M149; LOC100620468; MGC149895; MGC88244; pentraxin 1; PTX1; RP11-419N10.4 | |
Crp | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.