Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588023 Supplier Invitrogen™ Supplier No. PA588023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPNWGGGKKC GVCQKTVYFA EEVQCEGNSF HKSCFLCMVC KKNLDSTTVA VHGEEIYCKS CYGKKYGPKG YGYGQGAGTL STDKGESLGI KHEEAPGHRP TTNPNASKFA QKIGGSERCP RCSQAVYAAE KVIGAGKSWH KACFRCAKCG KGLESTTLAD KDGEIYCKGC YAKNFGPKGF GFGQGAGALV HSE; Positive Samples: A-549, BT-474, HeLa, Mouse lung, Mouse brain; Cellular Location: Nucleus.

This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRP1
Applications ELISA, Immunoprecipitation, Western Blot, RNA Immunoprecipitation
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CSRP1
Gene Accession No. P21291, P97315
Gene Alias AA408841; AA959891; AW545626; CRP; CRP1; CSRP; Csrp 1; Csrp1; CYRP; cysteine and glycine rich protein 1; cysteine and glycine-rich protein 1; cysteine rich protein 1; cysteine-rich protein 1; D1S181E; Epididymis luminal protein 141; epididymis secretory protein Li 286; HEL141; HEL-141; HEL-S-286; LIM-domain protein
Gene Symbols CSRP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human CSRP1 (NP_0040691).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13007, 1465
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.