Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118824 Supplier Invitrogen™ Supplier No. PA5118824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CSRP1.

This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSRP1
Gene Accession No. P21291, P47875, P97315
Gene Alias AA408841; AA959891; AW545626; CRP; CRP1; CSRP; Csrp 1; Csrp1; CYRP; cysteine and glycine rich protein 1; cysteine and glycine-rich protein 1; cysteine rich protein 1; cysteine-rich protein 1; D1S181E; Epididymis luminal protein 141; epididymis secretory protein Li 286; HEL141; HEL-141; HEL-S-286; LIM-domain protein
Gene Symbols CSRP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSRP1(Accession P21291), corresponding to amino acid residues C58-K108.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13007, 1465, 29276
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.