Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529999
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529999 Supplier Invitrogen™ Supplier No. PA529999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HCT116. Predicted reactivity: Mouse (99%), Rat (98%), Xenopus laevis (80%), Chicken (89%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the cysteine-rich protein (CSRP) family. This gene family includes a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this gene product occurs in proteins with critical functions in gene regulation, cell growth, and somatic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CSRP1
Gene Accession No. P21291
Gene Alias AA408841; AA959891; AW545626; CRP; CRP1; CSRP; Csrp 1; Csrp1; CYRP; cysteine and glycine rich protein 1; cysteine and glycine-rich protein 1; cysteine rich protein 1; cysteine-rich protein 1; D1S181E; Epididymis luminal protein 141; epididymis secretory protein Li 286; HEL141; HEL-141; HEL-S-286; LIM-domain protein
Gene Symbols CSRP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CSRP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.