Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100457 Supplier Invitrogen™ Supplier No. PA5100457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CSRP2.

CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSRP2
Gene Accession No. P97314, Q16527, Q62908
Gene Alias AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein
Gene Symbols CSRP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSRP2(Accession Q16527), corresponding to amino acid residues N81-K131.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13008, 1466, 29317
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.